accessibility menu, dialog, popup

UserName

CH3NNH3CI

CAS RN 65039-05-6

IUPAC Name: 1-butyl-3-methyl-1H-imidazol-3-ium iodide
Synonyms: 1-butyl-3-methylimidazolium iodide
Molecular Weight (g/mol): 266.13
Molecular Formula: C8H15IN2
InChi Key: XREPTGNZZKNFQZ-UHFFFAOYSA-M
SMILES: [I-].CCCCN1C=C[N+](C)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results