accessibility menu, dialog, popup

UserName

H3CS

CAS RN 65016-63-9

IUPAC Name: 3-nonylthiophene
Synonyms: 3-nonylthiophene
Molecular Weight (g/mol): 210.38
Molecular Formula: C13H22S
InChi Key: UUHSVAMCIZLNDQ-UHFFFAOYSA-N
SMILES: CCCCCCCCCC1=CSC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results