CAS RN 65-31-6
CAS RN 65-31-6
IUPAC Name:
bis((2R,3R)-2,3-dihydroxybutanedioic acid); 3-(1-methylpyrrolidin-2-yl)pyridine
Synonyms:
bis(L(+)-tartaric acid); nicotine
Molecular Weight (g/mol):
462.41
Molecular Formula:
C18H26N2O12
InChi Key:
RFEJUZJILGIRHQ-WBPXWQEISA-N
SMILES:
O[C@H]([C@@H](O)C(O)=O)C(O)=O.O[C@H]([C@@H](O)C(O)=O)C(O)=O.CN1CCCC1C1=CC=CN=C1
1
–
1
of
1
results
1
phyproof™ Nicotine ditartrate, ≥95% (HPLC), MilliporeSigma™ Supelco™
This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG