CAS RN 64741-65-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Neo-Clear™, (Xylene Substitute), For Microscopy, MilliporeSigma™
CAS: 64741-65-7