accessibility menu, dialog, popup

UserName

ONOOHNSHNCl

CAS RN 6470-87-7

IUPAC Name: 1-(4-chlorophenyl)-3-(2-nitrobenzoyl)thiourea
Synonyms: 1-(4-chlorophenyl)-3-(2-nitrobenzoyl)thiourea
Molecular Weight (g/mol): 335.76
Molecular Formula: C14H10ClN3O3S
InChi Key: QBUULMJMEMIBIA-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=CC=C1C(=O)NC(=S)NC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results