CAS RN 64559-06-4
IUPAC Name:
3-methoxybenzene-1-carbothioamide
Synonyms:
3-methoxybenzenecarbothioamide
Molecular Weight (g/mol):
167.23
Molecular Formula:
C8H9NOS
InChi Key:
LQSZSWBMLMGWPC-UHFFFAOYSA-N
SMILES:
COC1=CC=CC(=C1)C(N)=S
1
–
1
of
1
results
1
Promotions Available
3-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals
3-Methoxythiobenzamide, 97%, Thermo Scientific Chemicals
CAS: 64559-06-4 Molecular Formula: C8H9NOS Molecular Weight (g/mol): 167.226 MDL Number: MFCD04627361 InChI Key: LQSZSWBMLMGWPC-UHFFFAOYSA-N Synonym: 3-methoxythiobenzamide,3-methoxybenzothioamide,3-methoxy-thiobenzamide,benzenecarbothioamide,3-methoxy,amino 3-methoxyphenyl methane-1-thione,3-methoxybenzene-1-carbothioamide,acmc-20an1d,#,benzenecarbothioamide, 3-methoxy PubChem CID: 2060838 IUPAC Name: 3-methoxybenzenecarbothioamide SMILES: COC1=CC=CC(=C1)C(=S)N