accessibility menu, dialog, popup

UserName

CAS RN 64550-71-6

H3COOOSH3COOH3COOCH3OOCH3

CAS RN 64550-71-6

IUPAC Name: [3,4,5-tris(acetyloxy)-6-(methylsulfanyl)oxan-2-yl]methyl acetate
Synonyms: [3,4,5-tris(acetyloxy)-6-(methylsulfanyl)oxan-2-yl]methyl acetate
Molecular Weight (g/mol): 378.39
Molecular Formula: C15H22O9S
InChi Key: XWFUCHLBRWBKGN-UHFFFAOYNA-N
SMILES: CSC1OC(COC(C)=O)C(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results