CAS RN 64372-62-9
IUPAC Name:
[2-chloro-5-(trifluoromethyl)phenyl]methanol
Synonyms:
[2-chloro-5-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol):
210.58
Molecular Formula:
C8H6ClF3O
InChi Key:
VEKMDPQNERMQMO-UHFFFAOYSA-N
SMILES:
OCC1=CC(=CC=C1Cl)C(F)(F)F
1
–
1
of
1
results
1
2-Chloro-5-(trifluoromethyl)benzyl Alcohol 98.0+%, TCI America™
CAS: 64372-62-9 Molecular Formula: C8H6ClF3O Molecular Weight (g/mol): 210.58 MDL Number: MFCD01631469 InChI Key: VEKMDPQNERMQMO-UHFFFAOYSA-N Synonym: 2-chloro-5-trifluoromethyl benzyl alcohol,2-chloro-5-trifluoromethylbenzyl alcohol,2-chloro-5-trifluoromethyl phenyl methanol,2-chloro-5-trifluoromethylbenzylalcohol,2-chloro-5-trifluoromethyl benzylalcohol,4-chloro-3-hydroxymethyl benzotrifluoride,2-chloro-5-trifluoromethyl benzenemethanol,benzenemethanol, 2-chloro-5-trifluoromethyl,2-chloro-5-trifluoromethyl phenyl methan-1-ol PubChem CID: 2778110 IUPAC Name: [2-chloro-5-(trifluoromethyl)phenyl]methanol SMILES: C1=CC(=C(C=C1C(F)(F)F)CO)Cl