accessibility menu, dialog, popup

UserName

NaH3CONClOHN(R)(R)SCH3CH3(S)NOOO

CAS RN 642-78-4

IUPAC Name: sodium (2S,5R,6R)-6-[3-(2-chlorophenyl)-5-methyl-1,2-oxazole-4-amido]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
Synonyms: cloxacillin sodium anhydrous monosodium cloxacillin
Molecular Weight (g/mol): 457.86
Molecular Formula: C19H17ClN3NaO5S
InChi Key: SCLZRKVZRBKZCR-SLINCCQESA-M
SMILES: [Na+].CC1=C(C(=O)N[C@H]2[C@H]3SC(C)(C)[C@@H](N3C2=O)C([O-])=O)C(=NO1)C1=CC=CC=C1Cl
Color 
  • (1)
Quantity 
  • (1)
Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results