accessibility menu, dialog, popup

UserName

CAS RN 64169-67-1

NOF

CAS RN 64169-67-1

IUPAC Name: 1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile
Synonyms: 1-(4-fluorophenyl)-1,3-dihydro-2-benzofuran-5-carbonitrile
Molecular Weight (g/mol): 239.25
Molecular Formula: C15H10FNO
InChi Key: YXCRMKYHFFMNPT-UHFFFAOYNA-N
SMILES: FC1=CC=C(C=C1)C1OCC2=C1C=CC(=C2)C#N
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results