accessibility menu, dialog, popup

UserName

OPNNN

CAS RN 6415-07-2

IUPAC Name: 1-[bis(pyrrolidin-1-yl)phosphoroso]pyrrolidine
Synonyms: 1-[bis(pyrrolidin-1-yl)phosphoroso]pyrrolidine
Molecular Weight (g/mol): 257.32
Molecular Formula: C12H24N3OP
InChi Key: GQOGEHIVQIMJMO-UHFFFAOYSA-N
SMILES: O=P(N1CCCC1)(N1CCCC1)N1CCCC1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results