accessibility menu, dialog, popup

UserName

CAS RN 64138-65-4

IUPAC Name: 5-nitro-N-(1-phenylethyl)pyridin-2-amine
Synonyms: 5-nitro-N-(1-phenylethyl)pyridin-2-amine
Molecular Weight (g/mol): 243.27
Molecular Formula: C13H13N3O2
InChi Key: RAKMWGKBCDCUDX-UHFFFAOYNA-N
SMILES: CC(NC1=NC=C(C=C1)[N+]([O-])=O)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results