accessibility menu, dialog, popup

UserName

CAS RN 64010-42-0

IUPAC Name: magnesium(2+) hydrate diperchlorate
Synonyms: magnesium(2+) hydrate diperchlorate
Molecular Weight (g/mol): 241.21
Molecular Formula: Cl2H2MgO9
InChi Key: KKFFNQXCJMNXHI-UHFFFAOYSA-L
SMILES: O.[Mg++].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results