accessibility menu, dialog, popup

UserName

NaNaOClClSClClO

CAS RN 6385-58-6

IUPAC Name: disodium 2,4-dichloro-6-[(3,5-dichloro-2-oxidophenyl)sulfanyl]benzen-1-olate
Synonyms: disodium 2,4-dichloro-6-[(3,5-dichloro-2-oxidophenyl)sulfanyl]benzenolate
Molecular Weight (g/mol): 400
Molecular Formula: C12H4Cl4Na2O2S
InChi Key: FNYZFZRGBBCWBI-UHFFFAOYSA-L
SMILES: [Na+].[Na+].[O-]C1=C(Cl)C=C(Cl)C=C1SC1=CC(Cl)=CC(Cl)=C1[O-]
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results