accessibility menu, dialog, popup

UserName

HClCH3H2N

CAS RN 638-03-9

IUPAC Name: hydrogen 3-methylaniline chloride
Synonyms: hydrogen M-toluidine chloride
Molecular Weight (g/mol): 143.61
Molecular Formula: C7H10ClN
InChi Key: VDXZPXIEEVOGLV-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CC1=CC=CC(N)=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results