accessibility menu, dialog, popup

UserName

CAS RN 6376-59-6

H3CCH3NHNOOHOHHO(E)

CAS RN 6376-59-6

IUPAC Name: 3,4,5-trihydroxy-N'-[(E)-[4-(propan-2-yl)phenyl]methylidene]benzohydrazide
Synonyms: 3,4,5-trihydroxy-N'-[(E)-(4-isopropylphenyl)methylidene]benzohydrazide
Molecular Weight (g/mol): 314.34
Molecular Formula: C17H18N2O4
InChi Key: WSJPQOZRXXZSMG-GIJQJNRQSA-N
SMILES: CC(C)C1=CC=C(\C=N\NC(=O)C2=CC(O)=C(O)C(O)=C2)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results