accessibility menu, dialog, popup

UserName

H3CNCH3NHNCH3H3C

CAS RN 637-31-0

IUPAC Name: N4-[4-(dimethylamino)phenyl]-N1,N1-dimethylbenzene-1,4-diamine
Synonyms: N4-[4-(dimethylamino)phenyl]-N1,N1-dimethylbenzene-1,4-diamine
Molecular Weight (g/mol): 255.37
Molecular Formula: C16H21N3
InChi Key: ZPGZNKGKCSYBGK-UHFFFAOYSA-N
SMILES: CN(C)C1=CC=C(NC2=CC=C(C=C2)N(C)C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results