CAS RN 6362-63-6
IUPAC Name:
ethyl 3-cyano-2-oxo-3-phenylpropanoate
Synonyms:
ethyl 3-cyano-2-oxo-3-phenylpropanoate
Molecular Weight (g/mol):
217.22
Molecular Formula:
C12H11NO3
InChi Key:
YWNRATDDUSMBPR-UHFFFAOYNA-N
SMILES:
CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1
1
–
1
of
1
results
1
Promotions Available
Ethyl 3-cyano-3-phenylpyruvate, 97%
Ethyl 3-cyano-3-phenylpyruvate, 97%
CAS: 6362-63-6 Molecular Formula: C12H11NO3 Molecular Weight (g/mol): 217.22 MDL Number: MFCD00019801 InChI Key: YWNRATDDUSMBPR-UHFFFAOYNA-N Synonym: ethyl 3-cyano-3-phenylpyruvate,ethyl phenylcyanopyruvate,cyanophenylpyruvic acid ethyl ester,ethyl cyanophenylpyruvate,ethyl alpha-cyanophenylpyruvate,pyruvic acid, cyanophenyl-, ethyl ester,ethyl .alpha.-cyanophenylpyruvate,wln: ncr x1v1uno2,4-10-00-03440 beilstein handbook reference,benzenepropanoic acid, .beta.-cyano-.alpha.-oxo-, ethyl ester PubChem CID: 22845 IUPAC Name: ethyl 3-cyano-2-oxo-3-phenylpropanoate SMILES: CCOC(=O)C(=O)C(C#N)C1=CC=CC=C1