accessibility menu, dialog, popup

UserName

CAS RN 6351-10-6

HO

CAS RN 6351-10-6

IUPAC Name: 2,3-dihydro-1H-inden-1-ol
Synonyms: indanol (+-)-1-indanol 1-indanol, (+-)- (s)-1-indanol (S)-1-indanol (r)-1-indanol (R)-1-indanol 1-indanol, (S)- 1-indanol, (s)- 1-indanol, (R)-
Molecular Weight (g/mol): 134.18
Molecular Formula: C9H10O
InChi Key: YIAPLDFPUUJILH-UHFFFAOYNA-N
SMILES: OC1CCC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)