accessibility menu, dialog, popup

UserName

CAS RN 635-78-9

IUPAC Name: 7-hydroxy-3H-phenoxazin-3-one
Synonyms: resorufin resorufine
Molecular Weight (g/mol): 213.19
Molecular Formula: C12H7NO3
InChi Key: HSSLDCABUXLXKM-UHFFFAOYSA-N
SMILES: OC1=CC=C2N=C3C=CC(=O)C=C3OC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results