CAS RN 6342-70-7
IUPAC Name:
methyl 2-hydroxy-3-methoxybenzoate
Synonyms:
methyl 2-hydroxy-3-methoxybenzoate
Molecular Weight (g/mol):
182.18
Molecular Formula:
C9H10O4
InChi Key:
BWRCJLJJIXYLNV-UHFFFAOYSA-N
SMILES:
COC(=O)C1=CC=CC(OC)=C1O
1
–
1
of
1
results
1
Methyl 3-Methoxysalicylate 98.0+%, TCI America™
CAS: 6342-70-7 Molecular Formula: C9H10O4 Molecular Weight (g/mol): 182.175 MDL Number: MFCD00239511 InChI Key: BWRCJLJJIXYLNV-UHFFFAOYSA-N Synonym: 3-Methoxysalicylic Acid Methyl Ester, Methyl 2-Hydroxy-3-methoxybenzoate, 2-Hydroxy-3-methoxybenzoic Acid Methyl Ester, Methyl 2-Hydroxy-m-anisate, 2-Hydroxy-m-anisic Acid Methyl Ester PubChem CID: 80651 IUPAC Name: methyl 2-hydroxy-3-methoxybenzoate SMILES: COC1=CC=CC(=C1O)C(=O)OC