accessibility menu, dialog, popup

UserName

CAS RN 62999-73-9

IUPAC Name: diethyl (1,3-dithian-2-yl)phosphonate
Synonyms: diethyl 1,3-dithian-2-ylphosphonate
Molecular Weight (g/mol): 256.31
Molecular Formula: C8H17O3PS2
InChi Key: BGXBJZCFBVTFFP-UHFFFAOYSA-N
SMILES: CCOP(=O)(OCC)C1SCCCS1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results