accessibility menu, dialog, popup

UserName

CH3OO(R)(R)OCH3OOOO

CAS RN 6283-74-5

IUPAC Name: (3R,4R)-4-(acetyloxy)-2,5-dioxooxolan-3-yl acetate
Synonyms: L-diacetyltartaric anhydride
Molecular Weight (g/mol): 216.15
Molecular Formula: C8H8O7
InChi Key: XAKITKDHDMPGPW-IOMOGOHMNA-N
SMILES: CC(=O)O[C@@H]1[C@@H](OC(C)=O)C(=O)OC1=O
Percent Purity 
  • (1)
  • (2)
  • (1)
Quantity 
  • (1)
  • (2)
  • (1)
Physical Form 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)