accessibility menu, dialog, popup

UserName

BOOH3CCH3H3CH3CONCH3CH3

CAS RN 627899-90-5

IUPAC Name: dimethyl({3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propyl})amine
Synonyms: dimethyl({3-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenoxy]propyl})amine
Molecular Weight (g/mol): 305.23
Molecular Formula: C17H28BNO3
InChi Key: ONRJKPSMQGEPHV-UHFFFAOYSA-N
SMILES: CN(C)CCCOC1=CC=C(C=C1)B1OC(C)(C)C(C)(C)O1
Molecular Weight (g/mol) 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results