accessibility menu, dialog, popup

UserName

CAS RN 627-20-3

IUPAC Name: (2Z)-pent-2-ene
Synonyms: cis-2-pentene
Molecular Weight (g/mol): 70.14
Molecular Formula: C5H10
InChi Key: QMMOXUPEWRXHJS-HYXAFXHYSA-N
SMILES: CC\C=C/C
Boiling Point 
  • (1)
  • (3)
Percent Purity 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (3)
  • (1)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)