accessibility menu, dialog, popup

UserName

CAS RN 62625-17-6

KOSOOClClCl

CAS RN 62625-17-6

IUPAC Name: potassium 2,4,5-trichlorobenzene-1-sulfonate
Synonyms: potassium 2,4,5-trichlorobenzenesulfonate
Molecular Weight (g/mol): 299.59
Molecular Formula: C6H2Cl3KO3S
InChi Key: DRPLRPSXUUSFOB-UHFFFAOYSA-M
SMILES: [K+].[O-]S(=O)(=O)C1=CC(Cl)=C(Cl)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results