accessibility menu, dialog, popup

UserName

H3COOHNNH2O

CAS RN 626-36-8

IUPAC Name: ethyl N-carbamoylcarbamate
Synonyms: ethyl N-carbamoylcarbamate
Molecular Weight (g/mol): 132.12
Molecular Formula: C4H8N2O3
InChi Key: PIHPSKJRLDSJPX-UHFFFAOYSA-N
SMILES: CCOC(=O)NC(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results