accessibility menu, dialog, popup

UserName

CAS RN 6257-49-4

IUPAC Name: 2-(benzylamino)butan-1-ol
Synonyms: 2-(benzylamino)butan-1-ol
Molecular Weight (g/mol): 179.26
Molecular Formula: C11H17NO
InChi Key: PGFBTQBTIYCCFJ-UHFFFAOYNA-N
SMILES: CCC(CO)NCC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results