accessibility menu, dialog, popup

UserName

CH3H3CH3CH3C

CAS RN 625-65-0

IUPAC Name: 2,4-dimethylpent-2-ene
Synonyms: 2-pentene, 2,4-dimethyl-
Molecular Weight (g/mol): 98.19
Molecular Formula: C7H14
InChi Key: VVCFYASOGFVJFN-UHFFFAOYSA-N
SMILES: CC(C)C=C(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results