accessibility menu, dialog, popup

UserName

NH2ClNCl

CAS RN 62476-56-6

IUPAC Name: 2,6-dichloropyridin-3-amine
Synonyms: 3-amino-2,6-dichloropyridine
Molecular Weight (g/mol): 163
Molecular Formula: C5H4Cl2N2
InChi Key: MJVZSRZTBDMYLX-UHFFFAOYSA-N
SMILES: NC1=CC=C(Cl)N=C1Cl
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)