CAS RN 62451-84-7
IUPAC Name:
methyl 2-[3-(trifluoromethyl)phenyl]acetate
Synonyms:
methyl 2-[3-(trifluoromethyl)phenyl]acetate
Molecular Weight (g/mol):
218.18
Molecular Formula:
C10H9F3O2
InChi Key:
GMEBDKGPPKAPEM-UHFFFAOYSA-N
SMILES:
COC(=O)CC1=CC=CC(=C1)C(F)(F)F
1
–
2
of
2
results
1
Promotions Available
Methyl 3-(trifluoromethyl)phenylacetate, 95%
Methyl 3-(trifluoromethyl)phenylacetate, 95%
CAS: 62451-84-7 Molecular Formula: C10H9F3O2 Molecular Weight (g/mol): 218.175 MDL Number: MFCD04039762 InChI Key: GMEBDKGPPKAPEM-UHFFFAOYSA-N PubChem CID: 2734603 IUPAC Name: methyl 2-[3-(trifluoromethyl)phenyl]acetate SMILES: COC(=O)CC1=CC(=CC=C1)C(F)(F)F
2
Promotions Available
Methyl 3-(trifluoromethyl)phenylacetate, 97%, Thermo Scientific™