accessibility menu, dialog, popup

UserName

OHFOCl

CAS RN 62433-26-5

IUPAC Name: 2-(4-chlorobenzoyl)-4-fluorophenol
Synonyms: 2-(4-chlorobenzoyl)-4-fluorophenol
Molecular Weight (g/mol): 250.65
Molecular Formula: C13H8ClFO2
InChi Key: AYBQWBCUAWOLCT-UHFFFAOYSA-N
SMILES: OC1=CC=C(F)C=C1C(=O)C1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results