accessibility menu, dialog, popup

UserName

CAS RN 624-41-9

H3CCH3OCH3O

CAS RN 624-41-9

IUPAC Name: 2-methylbutyl acetate
Synonyms: β-methylbutyl acetate
Molecular Weight (g/mol): 130.19
Molecular Formula: C7H14O2
InChi Key: XHIUFYZDQBSEMF-UHFFFAOYNA-N
SMILES: CCC(C)COC(C)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results