accessibility menu, dialog, popup

UserName

CAS RN 624-16-8

H3COCH3

CAS RN 624-16-8

IUPAC Name: decan-4-one
Synonyms: 4-decanone
Molecular Weight (g/mol): 156.27
Molecular Formula: C10H20O
InChi Key: MKJDUHZPLQYUCB-UHFFFAOYSA-N
SMILES: CCCCCCC(=O)CCC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results