accessibility menu, dialog, popup

UserName

CAS RN 623-04-1

IUPAC Name: (4-aminophenyl)methanol
Synonyms: P-aminobenzylalcohol
Molecular Weight (g/mol): 123.16
Molecular Formula: C7H9NO
InChi Key: AXKGIPZJYUNAIW-UHFFFAOYSA-N
SMILES: NC1=CC=C(CO)C=C1
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (2)
  • (3)
  • (1)
Physical Form 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (4)
  • (2)
  • (3)
  • (1)
  • (4)
  • (2)