accessibility menu, dialog, popup

UserName

CAS RN 6223-35-4

IUPAC Name: sodium 3,8-dimethyl-5-(propan-2-yl)azulene-1-sulfonate
Synonyms: sodium guaiazulene sulfonate
Molecular Weight (g/mol): 300.35
Molecular Formula: C15H17NaO3S
InChi Key: GEYJUFBPCGDENK-UHFFFAOYSA-M
SMILES: [Na+].CC(C)C1=CC=C(C)C2=C(C=C(C)C2=C1)S([O-])(=O)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results