accessibility menu, dialog, popup

UserName

CAS RN 62162-97-4

Br

CAS RN 62162-97-4

IUPAC Name: 2-bromophenanthrene
Synonyms: 2-bromophenanthrene
Molecular Weight (g/mol): 257.13
Molecular Formula: C14H9Br
InChi Key: SQTPFYJEKHTINP-UHFFFAOYSA-N
SMILES: BrC1=CC=C2C(C=CC3=CC=CC=C23)=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results