CAS RN 6209-17-2
IUPAC Name:
N-(4-aminobenzenesulfonyl)-N-sodioacetamide
Synonyms:
N-(4-aminobenzenesulfonyl)-N-sodioacetamide
Molecular Weight (g/mol):
236.22
Molecular Formula:
C8H9N2NaO3S
InChi Key:
PQMSFAORUFMASU-UHFFFAOYSA-M
SMILES:
CC(=O)N([Na])S(=O)(=O)C1=CC=C(N)C=C1
1
–
1
of
1
results
1
Sulfacetamide Sodium Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Sulfacetamide Sodium belongs to the class of sulfonamides, widely used in ophthalmic preparations, to treat different ocular infections such as corneal ulcers and conjunctivitis. It shows bactericidal activity against susceptible microorganisms.