accessibility menu, dialog, popup

UserName

CAS RN 619-05-6

IUPAC Name: 3,4-diaminobenzoate
Synonyms: 3,4-diaminobenzoate
Molecular Weight (g/mol): 151.15
Molecular Formula: C7H7N2O2
InChi Key: HEMGYNNCNNODNX-UHFFFAOYSA-M
SMILES: NC1=CC=C(C=C1N)C([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (4)
Quantity 
  • (2)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (1)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (1)
  • (6)
  • (3)
  • (4)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (3)
  • (1)
  • (3)