CAS RN 615-45-2
IUPAC Name:
dihydrogen 2-methylbenzene-1,4-diamine dichloride
Synonyms:
2,5-diaminotoluene
Molecular Weight (g/mol):
195.09
Molecular Formula:
C7H12Cl2N2
InChi Key:
VQUHVWVGRKTIBH-UHFFFAOYSA-N
SMILES:
[H+].[H+].[Cl-].[Cl-].CC1=CC(N)=CC=C1N
1
–
1
of
1
results
1
2,5-Diaminotoluene Dihydrochloride 98.0+%, TCI America™
CAS: 615-45-2 Molecular Formula: C7H12Cl2N2 Molecular Weight (g/mol): 195.09 MDL Number: MFCD00035512 InChI Key: VQUHVWVGRKTIBH-UHFFFAOYSA-N Synonym: 2,5-Toluenediamine Dihydrochloride, Tolylene-2,5-diamine Dihydrochloride, 2-Methyl-1,4-phenylenediamine Dihydrochloride PubChem CID: 11996 IUPAC Name: dihydrogen 2-methylbenzene-1,4-diamine dichloride SMILES: [H+].[H+].[Cl-].[Cl-].CC1=CC(N)=CC=C1N