accessibility menu, dialog, popup

UserName

H3CO(S)O(S)H3COCH3OHHNO(S)CH3(S)OH(R)CH3(R)OH(R)CH3(S)OCH3O(R)CH3CH3NHNNOOOH(?)(Z)(Z)(E)

CAS RN 61379-65-5

IUPAC Name: (7S,9E,11S,12R,13S,14R,15R,16R,17S,18S,19E,21Z,26E)-26-{[(4-cyclopentylpiperazin-1-yl)amino]methylidene}-2,15,17,29-tetrahydroxy-11-methoxy-3,7,12,14,16,18,22-heptamethyl-6,23,27-trioxo-8,30-dioxa-24-azatetracyclo[23.3.1.1⁴,⁷.0⁵,²⁸]triaconta-1,3,5(28),9,19,21,25(29)-heptaen-13-yl acetate
Synonyms: rifapentine
Molecular Weight (g/mol): 877.05
Molecular Formula: C47H64N4O12
InChi Key: RISDZDVHYCURHD-QLWTZYDESA-N
SMILES: CO[C@H]1\C=C\O[C@@]2(C)OC3=C(C)C(O)=C4C(O)=C(NC(=O)\C(C)=C/C=C/[C@H](C)[C@H](O)[C@@H](C)[C@@H](O)[C@@H](C)[C@H](OC(C)=O)[C@@H]1C)\C(=C/NN1CCN(CC1)C1CCCC1)C(=O)C4=C3C2=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results