accessibility menu, dialog, popup

UserName

H3COOONOO

CAS RN 61312-84-3

IUPAC Name: (4-nitrophenyl)methyl 3-oxobutanoate
Synonyms: (4-nitrophenyl)methyl 3-oxobutanoate
Molecular Weight (g/mol): 237.21
Molecular Formula: C11H11NO5
InChi Key: KQPGVCMZXFBYMM-UHFFFAOYSA-N
SMILES: CC(=O)CC(=O)OCC1=CC=C(C=C1)[N+]([O-])=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results