accessibility menu, dialog, popup

UserName

CAS RN 613-08-1

IUPAC Name: anthracene-2-carboxylic acid
Synonyms: 2-anthracenecarboxylic acid
Molecular Weight (g/mol): 222.24
Molecular Formula: C15H10O2
InChi Key: RZRJYURCNBXIST-UHFFFAOYSA-N
SMILES: OC(=O)C1=CC=C2C=C3C=CC=CC3=CC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results