CAS RN 612845-44-0
CAS RN 612845-44-0
IUPAC Name:
(6-ethoxypyridin-3-yl)boronic acid
Synonyms:
6-ethoxypyridin-3-ylboronic acid
Molecular Weight (g/mol):
166.97
Molecular Formula:
C7H10BNO3
InChi Key:
UONCERAQKBPLML-UHFFFAOYSA-N
SMILES:
CCOC1=NC=C(C=C1)B(O)O