missing translation for 'onlineSavingsMsg'
Learn More

2-Ethoxypyridine-5-boronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. E11151G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E11151G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E11151G Supplier TCI America Supplier No. E11151G
Only null left

Specifications

Chemical Name or Material 2-Ethoxypyridine-5-boronic Acid (contains varying amounts of Anhydride)
CAS 612845-44-0
Melting Point 134°C
Color White
Molecular Formula C7H10BNO3
Quantity 1 g
MDL Number MFCD06201032
Synonym 6-ethoxypyridin-3-yl boronic acid, 2-ethoxy-5-pyridineboronic acid, 2-ethoxypyridine-5-boronic acid, 6-ethoxypyridine-3-boronic acid, 6-ethoxy-3-pyridinylboronic acid, 2-ethoxy-5-pyridylboronic acid, 3-borono-6-ethoxypyridine, 6-ethoxy-3-pyridylboronic acid, boronic acid, 6-ethoxy-3-pyridinyl, 6-ethoxy-3-pyridyl boronic acid
InChI Key UONCERAQKBPLML-UHFFFAOYSA-N
SMILES B(C1=CN=C(C=C1)OCC)(O)O
IUPAC Name (6-ethoxypyridin-3-yl)boronic acid
Molecular Weight (g/mol) 166.971
PubChem CID 12012237
Formula Weight 166.97
Physical Form Crystalline Powder
Show More Show Less