accessibility menu, dialog, popup

UserName

CAS RN 612-94-2

IUPAC Name: 2-phenylnaphthalene
Synonyms: naphthalene, 2-phenyl-
Molecular Weight (g/mol): 204.27
Molecular Formula: C16H12
InChi Key: TURIHPLQSRVWHU-UHFFFAOYSA-N
SMILES: C1=CC=C(C=C1)C1=CC=C2C=CC=CC2=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results