CAS RN 612-28-2
IUPAC Name:
N-methyl-2-nitroaniline
Synonyms:
N-methyl-O-nitroaniline
Molecular Weight (g/mol):
152.15
Molecular Formula:
C7H8N2O2
InChi Key:
KFBOUJZFFJDYTA-UHFFFAOYSA-N
SMILES:
CNC1=CC=CC=C1[N+]([O-])=O
1
–
1
of
1
results
1
Promotions Available
N-Methyl-2-nitroaniline, 98%
N-Methyl-2-nitroaniline, 98%
CAS: 612-28-2 Molecular Formula: C7H8N2O2 Molecular Weight (g/mol): 152.15 MDL Number: MFCD00007090 InChI Key: KFBOUJZFFJDYTA-UHFFFAOYSA-N Synonym: n-methyl-o-nitroaniline,benzenamine, n-methyl-2-nitro,n-methyl 2-nitroaniline,o-methylamino nitrobenzene,aniline, n-methyl-o-nitro,2-nitro-n-methylaniline,o-nitro-n-methylaniline,n-methyl-2-nitrobenzenamine,o-nitro-n-methyl aniline,methyl 2-nitrophenyl amine PubChem CID: 69157 IUPAC Name: N-methyl-2-nitroaniline SMILES: CNC1=CC=CC=C1[N+]([O-])=O