accessibility menu, dialog, popup

UserName

CAS RN 61-31-4

NaOO

CAS RN 61-31-4

IUPAC Name: sodium 2-(naphthalen-1-yl)acetate
Synonyms: sodium 2-naphthalen-1-ylacetate
Molecular Weight (g/mol): 208.19
Molecular Formula: C12H9NaO2
InChi Key: CJUUXVFWKYRHAR-UHFFFAOYSA-M
SMILES: [Na+].[O-]C(=O)CC1=C2C=CC=CC2=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results