accessibility menu, dialog, popup

UserName

CAS RN 6088-51-3

IUPAC Name: 6-[(6-hydroxynaphthalen-2-yl)disulfanyl]naphthalen-2-ol
Synonyms: ddd (analytical)
Molecular Weight (g/mol): 350.45
Molecular Formula: C20H14O2S2
InChi Key: AHXGXXJEEHFHDK-UHFFFAOYSA-N
SMILES: OC1=CC2=CC=C(SSC3=CC4=CC=C(O)C=C4C=C3)C=C2C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results