accessibility menu, dialog, popup

UserName

CH3OOHO

CAS RN 607-88-5

IUPAC Name: 4-methylphenyl 2-hydroxybenzoate
Synonyms: P-cresyl salicylate
Molecular Weight (g/mol): 228.25
Molecular Formula: C14H12O3
InChi Key: AMYKXJAXRJCLCV-UHFFFAOYSA-N
SMILES: CC1=CC=C(OC(=O)C2=CC=CC=C2O)C=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results